siRNA Details
Gene Symbol: PCNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268009.2 | XP_005268066.1 | 2 | 1402 | ||
GenBank mRNA | BC030714.1 | - | 1 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
