siRNA Details
Gene Symbol: TRAV12-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB305705.1 | - | 2 | 359 | ||
AB306068.1 | - | 2 | 351 | |||
AB306105.1 | - | 2 | 350 | |||
AB306116.1 | - | 2 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
