siRNA Details
Gene Symbol: REC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042367.1 | NP_001035826.1 | 3, 4 | 372 | ||
XM_017022083.1 | XP_016877572.1 | 2, 3 | 164 | |||
GenBank mRNA | AK131030.1 | - | 2, 3 | 282 | ||
BC055412.1 | - | 3, 4 | 353 | |||
BX113903.1 | - | 1, 2 | 83 | |||
DB465854.1 | - | 3, 4 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
