siRNA Details
Gene Symbol: LINC00528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103718.1 | 1 | 1803 | |||
GenBank mRNA | AK097861.1 | - | 1 | 1770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
