siRNA Details
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 5 | 681 | ||
XM_011515604.2 | XP_011513906.1 | 6 | 867 | |||
XM_011515608.2 | XP_011513910.1 | 6 | 771 | |||
XM_011515609.2 | XP_011513911.1 | 4 | 662 | |||
XM_011515610.2 | XP_011513912.1 | 7 | 948 | |||
XM_011515611.2 | XP_011513913.1 | 5 | 815 | |||
XM_017012777.1 | XP_016868266.1 | 4 | 516 | |||
XM_017012778.1 | XP_016868267.1 | 5 | 719 | |||
GenBank mRNA | AF277624.1 | - | 5 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
