siRNA Details
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 4, 5 | 533 | ||
XM_011515604.2 | XP_011513906.1 | 5, 6 | 719 | |||
XM_011515608.2 | XP_011513910.1 | 5, 6 | 623 | |||
XM_011515609.2 | XP_011513911.1 | 3, 4 | 514 | |||
XM_011515610.2 | XP_011513912.1 | 6, 7 | 800 | |||
XM_011515611.2 | XP_011513913.1 | 4, 5 | 667 | |||
XM_017012777.1 | XP_016868266.1 | 3, 4 | 368 | |||
XM_017012778.1 | XP_016868267.1 | 4, 5 | 571 | |||
GenBank mRNA | AF277624.1 | - | 4, 5 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
