siRNA Details
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321601.1 | NP_001308530.1 | 1 | 70 | ||
XM_017026804.1 | XP_016882293.1 | 1 | 197 | |||
XM_017026805.1 | XP_016882294.1 | 2 | 664 | |||
GenBank mRNA | BC157867.1 | - | 1 | 84 | ||
BC171868.1 | - | 1 | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
