siRNA Details
Gene Symbol: ZNF815P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023382.1 | 5 | 1280 | |||
GenBank mRNA | AK096288.1 | - | 5 | 1279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
