siRNA Details
Gene Symbol: GLTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014985.2 | NP_001014985.2 | 1 | 109 | ||
XM_005256626.3 | XP_005256683.1 | 1 | 152 | |||
GenBank mRNA | BC029290.1 | - | 1 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
