siRNA Details
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182530.2 | NP_872336.2 | 9, 10 | 1281 | ||
XM_011539052.2 | XP_011537354.1 | 8, 9 | 1762 | |||
XM_011539055.1 | XP_011537357.1 | 7, 8 | 749 | |||
XM_017018967.1 | XP_016874456.1 | 8, 9 | 1563 | |||
XM_017018968.1 | XP_016874457.1 | 9, 10 | 2017 | |||
XM_017018969.1 | XP_016874458.1 | 8, 9 | 1497 | |||
XM_017018970.1 | XP_016874459.1 | 7, 8 | 1307 | |||
XM_017018971.1 | XP_016874460.1 | 9, 10 | 2086 | |||
XM_017018972.1 | XP_016874461.1 | 9, 10 | 2085 | |||
XM_017018973.1 | XP_016874462.1 | 5, 6 | 978 | |||
XM_017018974.1 | XP_016874463.1 | 9, 10 | 2086 | |||
XM_017018975.1 | XP_016874464.1 | 9, 10 | 2086 | |||
XM_017018976.1 | XP_016874465.1 | 9, 10 | 2086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
