siRNA Details
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293083.1 | NP_001280012.1 | 43 | 4779 | ||
XM_011512110.2 | XP_011510412.1 | 43 | 4796 | |||
XM_011512111.2 | XP_011510413.1 | 43 | 4793 | |||
XM_011512113.2 | XP_011510415.1 | 43 | 4790 | |||
XM_011512114.2 | XP_011510416.1 | 43 | 4793 | |||
XM_011512115.2 | XP_011510417.1 | 42 | 4727 | |||
XM_011512116.2 | XP_011510418.1 | 43 | 4709 | |||
XM_011512117.2 | XP_011510419.1 | 42 | 4691 | |||
XM_011512118.2 | XP_011510420.1 | 42 | 4676 | |||
XM_011512119.2 | XP_011510421.1 | 42 | 4634 | |||
XM_011512120.2 | XP_011510422.1 | 42 | 4687 | |||
XM_011512122.2 | XP_011510424.1 | 43 | 4797 | |||
XM_011512124.2 | XP_011510426.1 | 27 | 4380 | |||
XM_017005222.1 | XP_016860711.1 | 43 | 4793 | |||
XM_017005223.1 | XP_016860712.1 | 26 | 3444 | |||
XM_017005224.1 | XP_016860713.1 | 17 | 2236 | |||
GenBank mRNA | AL133022.1 | - | 25 | 4182 | ||
AY461813.1 | - | 42 | 4805 | |||
BC117324.1 | - | 9 | 987 | |||
BC126368.1 | - | 9 | 990 | |||
BC143825.1 | - | 9 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
