siRNA Details
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293083.1 | NP_001280012.1 | 48, 49 | 5496 | ||
XM_011512110.2 | XP_011510412.1 | 48, 49 | 5540 | |||
XM_011512111.2 | XP_011510413.1 | 48, 49 | 5537 | |||
XM_011512113.2 | XP_011510415.1 | 48, 49 | 5534 | |||
XM_011512114.2 | XP_011510416.1 | 48, 49 | 5510 | |||
XM_011512115.2 | XP_011510417.1 | 47, 48 | 5471 | |||
XM_011512116.2 | XP_011510418.1 | 48, 49 | 5453 | |||
XM_011512117.2 | XP_011510419.1 | 47, 48 | 5435 | |||
XM_011512118.2 | XP_011510420.1 | 47, 48 | 5420 | |||
XM_011512119.2 | XP_011510421.1 | 47, 48 | 5378 | |||
XM_011512120.2 | XP_011510422.1 | 47, 48 | 5431 | |||
XM_011512122.2 | XP_011510424.1 | 48, 49 | 5541 | |||
XM_011512124.2 | XP_011510426.1 | 32, 33 | 5124 | |||
XM_017005222.1 | XP_016860711.1 | 48, 49 | 5537 | |||
XM_017005223.1 | XP_016860712.1 | 31, 32 | 4188 | |||
XM_017005224.1 | XP_016860713.1 | 22, 23 | 2980 | |||
GenBank mRNA | AL133022.1 | - | 30, 31 | 4899 | ||
AY461813.1 | - | 47, 48 | 5522 | |||
BC117324.1 | - | 14, 15 | 1704 | |||
BC126368.1 | - | 14, 15 | 1707 | |||
BC143825.1 | - | 14, 15 | 1704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
