siRNA Details
Gene Symbol: UBXN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077619.1 | NP_001071087.1 | 2, 3 | 308 | ||
XM_005251165.2 | XP_005251222.1 | 2, 3 | 211 | |||
XM_005251166.1 | XP_005251223.1 | 2, 3 | 211 | |||
GenBank mRNA | AK126300.1 | - | 2, 3 | 211 | ||
AL831990.2 | - | 2, 3 | 182 | |||
AL833075.2 | - | 2, 3 | 182 | |||
BC113645.1 | - | 2, 3 | 308 | 8322697 | ||
BX647876.2 | - | 2, 3 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
