siRNA Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080416.3 | NP_001073885.1 | 5, 6 | 927 | ||
NM_001144755.2 | NP_001138227.1 | 5, 6 | 927 | |||
NM_001294282.1 | NP_001281211.1 | 5, 6 | 927 | |||
XM_011517533.2 | XP_011515835.1 | 5, 6 | 968 | |||
XM_011517535.2 | XP_011515837.1 | 5, 6 | 968 | |||
XM_017013455.1 | XP_016868944.1 | 5, 6 | 778 | |||
XM_017013456.1 | XP_016868945.1 | 5, 6 | 775 | |||
XM_017013457.1 | XP_016868946.1 | 5, 6 | 776 | |||
XM_017013458.1 | XP_016868947.1 | 5, 6 | 773 | |||
XM_017013459.1 | XP_016868948.1 | 5, 6 | 780 | |||
GenBank mRNA | BC101186.1 | - | 5, 6 | 585 | 40018967 | |
BC101188.1 | - | 5, 6 | 585 | 40018969 | ||
X13269.1 | - | 5, 6 | 620 | |||
X13294.1 | - | 5, 6 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
