siRNA Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080416.3 | NP_001073885.1 | 12 | 2082 | ||
NM_001144755.2 | NP_001138227.1 | 12 | 2082 | |||
NM_001294282.1 | NP_001281211.1 | 12 | 2079 | |||
XM_011517533.2 | XP_011515835.1 | 12 | 2138 | |||
XM_011517535.2 | XP_011515837.1 | 12 | 2138 | |||
XM_017013455.1 | XP_016868944.1 | 12 | 1948 | |||
XM_017013456.1 | XP_016868945.1 | 12 | 1930 | |||
XM_017013457.1 | XP_016868946.1 | 12 | 1946 | |||
XM_017013458.1 | XP_016868947.1 | 12 | 1928 | |||
XM_017013459.1 | XP_016868948.1 | 12 | 1950 | |||
GenBank mRNA | AK308490.1 | - | 5 | 849 | ||
BC101186.1 | - | 12 | 1737 | 40018967 | ||
BC101188.1 | - | 12 | 1740 | 40018969 | ||
X13294.1 | - | 12 | 1775 | |||
X66087.1 | - | 3 | 482 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008651, NM_001290397, X82327, BC103534, BC103533, and others.
Gene Ontology Categories:
