siRNA Details
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 4 | 1396 | ||
XM_006723059.3 | XP_006723122.1 | 5 | 1442 | |||
XM_017026393.1 | XP_016881882.1 | 6 | 1687 | |||
XM_017026394.1 | XP_016881883.1 | 6 | 1612 | |||
XM_017026395.1 | XP_016881884.1 | 6 | 1760 | |||
XM_017026396.1 | XP_016881885.1 | 5 | 1529 | |||
XM_017026397.1 | XP_016881886.1 | 5 | 1534 | |||
XM_017026398.1 | XP_016881887.1 | 5 | 1413 | |||
XM_017026399.1 | XP_016881888.1 | 4 | 1406 | |||
XM_017026400.1 | XP_016881889.1 | 4 | 1326 | |||
GenBank mRNA | AK096348.1 | - | 1 | 62 | ||
AK129992.1 | - | 1 | 933 | |||
AL831902.1 | - | 1 | 594 | |||
BC105738.1 | - | 8 | 1708 | |||
BC131555.1 | - | 3 | 1251 | 40108031 | ||
CR933622.1 | - | 2 | 1182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
