siRNA Details
Gene Symbol: NCMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010980.4 | NP_001010980.1 | 4 | 885 | ||
XM_005245872.3 | XP_005245929.1 | 4 | 2223 | |||
XM_011541463.2 | XP_011539765.1 | 4 | 1682 | |||
GenBank mRNA | AK124519.1 | - | 4 | 850 | ||
BC050474.1 | - | 4 | 957 | |||
BC147014.1 | - | 3 | 806 | 9056942 | ||
BC147015.1 | - | 3 | 806 | 9056943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
