siRNA Details
Gene Symbol: FAM162B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085480.2 | NP_001078949.1 | 4 | 864 | ||
GenBank mRNA | BC038997.1 | - | 4 | 863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
