siRNA Details
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020877.3 | NP_065928.2 | 79 | 13644 | ||
XM_011523663.1 | XP_011521965.1 | 81 | 12512 | |||
XM_011523664.2 | XP_011521966.1 | 82 | 12661 | |||
XM_011523666.1 | XP_011521968.1 | 80 | 12323 | |||
XM_011523667.2 | XP_011521969.1 | 80 | 12415 | |||
XM_011523668.1 | XP_011521970.1 | 71 | 10895 | |||
XM_011523669.2 | XP_011521971.1 | 69 | 10579 | |||
XM_017024218.1 | XP_016879707.1 | 68 | 10596 | |||
GenBank mRNA | AB040936.2 | - | 79 | 13644 | ||
AK128517.1 | - | 20 | 3033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
