siRNA Details
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020877.3 | NP_065928.2 | 72 | 12439 | ||
XM_011523663.1 | XP_011521965.1 | 74 | 11307 | |||
XM_011523664.2 | XP_011521966.1 | 75 | 11456 | |||
XM_011523666.1 | XP_011521968.1 | 73 | 11118 | |||
XM_011523667.2 | XP_011521969.1 | 73 | 11210 | |||
XM_011523668.1 | XP_011521970.1 | 64 | 9690 | |||
XM_011523669.2 | XP_011521971.1 | 62 | 9374 | |||
XM_017024218.1 | XP_016879707.1 | 61 | 9391 | |||
XM_017024219.1 | XP_016879708.1 | 74 | 11307 | |||
GenBank mRNA | AB040936.2 | - | 72 | 12439 | ||
AK128517.1 | - | 13 | 1826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
