siRNA Details
Gene Symbol: PTGES2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024425.1 | 1 | 420 | |||
GenBank mRNA | AK094537.1 | - | 1 | 407 | ||
BC132745.1 | - | 1 | 378 | |||
BC132747.1 | - | 1 | 378 | |||
BC141829.1 | - | 1 | 367 | |||
BC151210.1 | - | 1 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
