siRNA Details
Gene Symbol: SYNDIG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105579.1 | NP_001099049.1 | 3 | 684 | ||
XM_017021600.1 | XP_016877089.1 | 3 | 3270 | |||
GenBank mRNA | DB173128.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
