siRNA Details
Gene Symbol: MUC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005961.2 | NP_005952.2 | 31 | 5514 | ||
GenBank mRNA | AK096772.1 | - | 1 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
