siRNA Details
Gene Symbol: AACSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024035.2 | 10 | 1353 | |||
NR_135095.1 | 10 | 1356 | ||||
GenBank mRNA | BC140764.1 | - | 10 | 1307 | 9021632 | |
BC144474.1 | - | 10 | 1306 | |||
DR008193.1 | - | 4 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
