siRNA Details
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 23 | 2472 | |||
GenBank mRNA | AL833251.1 | - | 23 | 2472 | ||
BC063633.1 | - | 21 | 2091 | |||
BC144350.1 | - | 4 | 447 | 9052870 | ||
KF175207.1 | - | 4 | 447 | |||
KF175220.1 | - | 4 | 352 | |||
KF175221.1 | - | 4 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
