siRNA Details
Gene Symbol: C15orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015492.4 | NP_056307.2 | 3 | 4155 | ||
GenBank mRNA | AF143881.1 | - | 1 | 643 | ||
AK057025.1 | - | 2 | 2082 | |||
AK125604.1 | - | 3 | 4124 | |||
AK128205.1 | - | 3 | 4245 | |||
AL117540.1 | - | 2 | 1214 | |||
AL834382.1 | - | 2 | 2301 | |||
BC000540.2 | - | 2 | 2345 | |||
BC011905.2 | - | 2 | 1303 | |||
BC110367.1 | - | 3 | 4142 | 5494248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
