siRNA Details
Gene Symbol: SLC38A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080442.2 | NP_001073911.1 | 7 | 821 | ||
XM_017022946.1 | XP_016878435.1 | 9 | 1286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
