siRNA Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277372.2 | NP_001264301.2 | 16, 17 | 2577 | ||
XM_006712172.2 | XP_006712235.1 | 16, 17 | 2582 | |||
XM_011510467.2 | XP_011508769.1 | 16, 17 | 2690 | |||
XM_011510468.2 | XP_011508770.1 | 16, 17 | 2585 | |||
GenBank mRNA | AB095932.1 | - | 4, 5 | 3085 | ||
BC118982.1 | - | 4, 5 | 898 | 40026312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
