siRNA Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277372.2 | NP_001264301.2 | 17, 18 | 2755 | ||
XM_006712172.2 | XP_006712235.1 | 17, 18 | 2760 | |||
XM_011510467.2 | XP_011508769.1 | 17, 18 | 2868 | |||
XM_011510468.2 | XP_011508770.1 | 17, 18 | 2763 | |||
GenBank mRNA | AB095932.1 | - | 5, 6 | 3263 | ||
BC118982.1 | - | 5, 6 | 1076 | 40026312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
