siRNA Details
Gene Symbol: TMEM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286487.1 | NP_001273416.1 | 19 | 2294 | ||
NM_015497.4 | NP_056312.2 | 20 | 2355 | |||
XM_005254287.1 | XP_005254344.1 | 20 | 2209 | |||
XM_006720468.2 | XP_006720531.1 | 20 | 2214 | |||
XM_011521443.2 | XP_011519745.1 | 19 | 2126 | |||
XM_017022067.1 | XP_016877556.1 | 20 | 2211 | |||
GenBank mRNA | AF129927.1 | - | 4 | 691 | ||
AF132733.1 | - | 20 | 2211 | |||
AI023965.1 | - | 1 | 81 | |||
AK074870.1 | - | 20 | 2204 | |||
AK075403.1 | - | 20 | 2241 | |||
AL049944.1 | - | 7 | 901 |
