siRNA Details
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024690.2 | NP_078966.2 | 1 | 1826 | ||
XM_017027486.1 | XP_016882975.1 | 4 | 1776 | |||
XM_017027487.1 | XP_016882976.1 | 4 | 1776 | |||
XM_017027488.1 | XP_016882977.1 | 4 | 1776 | |||
XM_017027489.1 | XP_016882978.1 | 1 | 1622 | |||
XM_017027490.1 | XP_016882979.1 | 4 | 1776 | |||
XM_017027491.1 | XP_016882980.1 | 4 | 1776 | |||
XM_017027492.1 | XP_016882981.1 | 4 | 1776 | |||
XM_017027493.1 | XP_016882982.1 | 4 | 1776 | |||
XM_017027494.1 | XP_016882983.1 | 4 | 1776 | |||
XM_017027495.1 | XP_016882984.1 | 4 | 1776 | |||
XM_017027496.1 | XP_016882985.1 | 4 | 1776 | |||
XM_017027497.1 | XP_016882986.1 | 4 | 1776 | |||
XM_017027498.1 | XP_016882987.1 | 4 | 1776 | |||
GenBank mRNA | AF414442.2 | - | 1826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
