siRNA Details
Gene Symbol: ZBTB7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039360.2 | NP_001034449.1 | 2 | 1178 | ||
NM_001318841.1 | NP_001305770.1 | 4 | 1359 | |||
XM_005258229.4 | XP_005258286.1 | 3 | 1103 | |||
XM_011525861.2 | XP_011524163.1 | 6 | 1583 | |||
XM_011525863.2 | XP_011524165.1 | 5 | 1700 | |||
XM_011525864.2 | XP_011524166.1 | 5 | 1455 | |||
XM_011525865.2 | XP_011524167.1 | 5 | 1696 | |||
XM_011525866.2 | XP_011524168.1 | 4 | 1441 | |||
XM_011525867.2 | XP_011524169.1 | 4 | 1230 | |||
XM_011525869.2 | XP_011524171.1 | 5 | 1705 | |||
XM_011525870.2 | XP_011524172.1 | 4 | 1401 | |||
XM_011525871.2 | XP_011524173.1 | 2 | 1916 | |||
XM_017025605.1 | XP_016881094.1 | 6 | 3378 | |||
XM_017025606.1 | XP_016881095.1 | 6 | 1832 | |||
XM_017025607.1 | XP_016881096.1 | 6 | 1587 | |||
XM_017025608.1 | XP_016881097.1 | 5 | 3255 | |||
XM_017025609.1 | XP_016881098.1 | 4 | 1572 | |||
GenBank mRNA | AM988836.1 | - | 2 | 1166 | ||
AM988837.1 | - | 2 | 1311 | |||
AM988838.1 | - | 3 | 1361 | |||
AM988839.1 | - | 4 | 1359 | |||
EF120357.1 | - | 2 | 1230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
