siRNA Details
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270989.1 | NP_001257918.1 | 4 | 462 | ||
XM_005265666.4 | XP_005265723.1 | 4 | 1334 | |||
XM_011531817.2 | XP_011530119.1 | 4 | 1320 | |||
GenBank mRNA | AK289455.1 | - | 4 | 460 | ||
AY358920.1 | - | 4 | 418 | |||
DQ235269.1 | - | 4 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
