siRNA Details
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270989.1 | NP_001257918.1 | 3 | 266 | ||
NM_001270990.1 | NP_001257919.1 | 3 | 266 | |||
NM_001270991.1 | NP_001257920.1 | 3 | 239 | |||
NM_001270992.1 | NP_001257921.1 | 3 | 266 | |||
NM_001270993.1 | NP_001257922.1 | 3 | 239 | |||
NR_073114.1 | 3 | 266 | ||||
NR_073117.1 | 2 | 176 | ||||
XM_005265666.4 | XP_005265723.1 | 3 | 1138 | |||
XM_011531817.2 | XP_011530119.1 | 3 | 1124 | |||
GenBank mRNA | AK289455.1 | - | 3 | 264 | ||
AY358920.1 | - | 3 | 222 | |||
BC127938.1 | - | 3 | 262 | 40132636 | ||
BC127939.1 | - | 3 | 262 | 40132642 | ||
CB150835.1 | - | 3 | 239 | |||
DQ235264.1 | - | 3 | 202 | |||
DQ235265.1 | - | 3 | 175 | |||
DQ235266.1 | - | 2 | 112 | |||
DQ235267.1 | - | 3 | 202 | |||
DQ235268.1 | - | 3 | 175 | |||
DQ235269.1 | - | 3 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
