siRNA Details
Gene Symbol: ROBO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290039.1 | NP_001276968.1 | 1 | 656 | ||
NM_001290040.1 | NP_001276969.1 | 1 | 656 | |||
NM_001290065.1 | NP_001276994.1 | 1 | 656 | |||
NM_002942.4 | NP_002933.1 | 1 | 656 | |||
XM_017006986.1 | XP_016862475.1 | 1 | 704 | |||
XM_017006993.1 | XP_016862482.1 | 1 | 704 | |||
XM_017006998.1 | XP_016862487.1 | 1 | 704 | |||
XM_017006999.1 | XP_016862488.1 | 1 | 704 | |||
XM_017007000.1 | XP_016862489.1 | 1 | 704 | |||
XM_017007006.1 | XP_016862495.1 | 1 | 704 | |||
GenBank mRNA | AB046788.1 | - | 1 | 19 | ||
BC144303.1 | - | 1 | 69 | |||
BC146772.1 | - | 1 | 19 | 9007219 | ||
DQ533874.1 | - | 1 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
