siRNA Details
Gene Symbol: C2CD4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136263.1 | NP_001129735.1 | 2 | 220 | ||
XM_011527694.1 | XP_011525996.1 | 2 | 178 | |||
GenBank mRNA | AB075837.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
