siRNA Details
Gene Symbol: LIPE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073179.1 | 1 | 1004 | |||
GenBank mRNA | AK096849.1 | - | 1 | 1004 | ||
AK310497.1 | - | 2 | 248 | |||
BC037865.2 | - | 1 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
