siRNA Details
Gene Symbol: SERPINE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101320.1 | NP_001094790.1 | 7 | 1171 | ||
XM_017020707.1 | XP_016876196.1 | 6 | 10131 | |||
GenBank mRNA | AM402969.1 | - | 7 | 1114 | ||
AX772926.1 | - | 7 | 1171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
