siRNA Details
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178310.3 | NP_840101.1 | 3 | 1219 | ||
GenBank mRNA | BC041461.1 | - | 3 | 1127 | ||
BC107058.2 | - | 3 | 1220 | 40005947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
