siRNA Details
Gene Symbol: C6orf120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 1057 | ||
NM_001317342.1 | NP_001304271.1 | 2 | 817 | |||
GenBank mRNA | AF055030.1 | - | 1 | 80 | ||
AF451991.1 | - | 1 | 1008 | |||
AK295061.1 | - | 1 | 1043 | |||
AK295962.1 | - | 2 | 792 | |||
BC051700.1 | - | 1 | 1008 | 4792634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
