siRNA Details
Gene Symbol: C6orf120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 1755 | ||
NM_001317342.1 | NP_001304271.1 | 2 | 1515 | |||
GenBank mRNA | AF055030.1 | - | 1 | 778 | ||
BC051700.1 | - | 1 | 1706 | 4792634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
