siRNA Details
Gene Symbol: MARCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102562.1 | NP_001096032.1 | 4 | 1341 | ||
GenBank mRNA | BC150513.1 | - | 2 | 470 | ||
BC150514.1 | - | 2 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
