siRNA Details
Gene Symbol: RPL32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003111.2 | 4 | 823 | |||
GenBank mRNA | BC015389.1 | - | 3 | 387 | ||
BC053996.1 | - | 5 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
