siRNA Details
Gene Symbol: BTNL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287262.1 | NP_001274191.1 | 2 | 385 | ||
XM_011544015.2 | XP_011542317.1 | 2 | 1593 | |||
XM_016999986.1 | XP_016855475.1 | 1 | 2315 | |||
XM_016999987.1 | XP_016855476.1 | 2 | 1593 | |||
GenBank mRNA | DY654633.1 | - | 2 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
