siRNA Details
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176819.3 | NP_789789.2 | 5 | 3502 | ||
XM_005272670.1 | XP_005272727.1 | 4 | 3303 | |||
XM_006724559.1 | XP_006724622.1 | 4 | 3225 | |||
GenBank mRNA | AF289581.1 | - | 1 | 754 | ||
AK055190.1 | - | 1 | 2154 | |||
AK056730.1 | - | 1 | 809 | |||
AK094014.1 | - | 1 | 1397 | |||
CR627166.1 | - | 1 | 1669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
