siRNA Details
Gene Symbol: DOC2GP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033791.1 | 2 | 112 | |||
GenBank mRNA | DY654453.1 | - | 4 | 365 | ||
DY654463.1 | - | 2 | 111 | |||
DY654465.1 | - | 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
