siRNA Details
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 5 | 564 | ||
XM_005273409.1 | XP_005273466.1 | 5 | 564 | |||
XM_005273410.1 | XP_005273467.1 | 5 | 564 | |||
XM_005273411.1 | XP_005273468.1 | 5 | 564 | |||
XM_005273412.3 | XP_005273469.1 | 5 | 564 | |||
XM_017013088.1 | XP_016868577.1 | 5 | 564 | |||
XM_017013089.1 | XP_016868578.1 | 5 | 468 | |||
GenBank mRNA | AK304441.1 | - | 5 | 516 | ||
BC042037.1 | - | 5 | 509 | |||
BE327420.1 | - | 4 | 412 | |||
DA395558.1 | - | 5 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
