siRNA Details
Gene Symbol: POLR2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015532.4 | NP_056347.1 | 2 | 765 | ||
GenBank mRNA | AF326773.1 | - | 2 | 455 | ||
AK074767.1 | - | 2 | 574 | |||
AK074955.1 | - | 2 | 569 | |||
AK223571.1 | - | 2 | 564 | |||
AK308873.1 | - | 2 | 745 | |||
AY339380.1 | - | 2 | 583 | |||
BC001510.2 | - | 2 | 555 | 3504557 | ||
BI833260.1 | - | 2 | 742 | |||
BQ072611.1 | - | 2 | 535 |
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285900.3 | NP_001272829.1 | 13 | 1765 | ||
NR_104371.3 | 15 | 2555 | ||||
GenBank mRNA | AY353058.1 | - | 15 | 2426 | ||
JF419331.1 | - | 13 | 1655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
