siRNA Details
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 9 | 6532 | ||
NM_032173.3 | NP_115549.2 | 9 | 6325 | |||
XM_011530435.2 | XP_011528737.1 | 10 | 6521 | |||
XM_011530436.2 | XP_011528738.1 | 11 | 6576 | |||
XM_011530437.1 | XP_011528739.1 | 10 | 7246 | |||
XM_011530438.2 | XP_011528740.1 | 10 | 6762 | |||
XM_017028990.1 | XP_016884479.1 | 10 | 7017 | |||
GenBank mRNA | AB051436.1 | - | 9 | 6398 | ||
AK022809.1 | - | 1 | 3148 | |||
BC042614.1 | - | 1 | 3361 | |||
BC051344.1 | - | 1 | 3453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
