siRNA Details
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 9 | 3420 | ||
NM_032173.3 | NP_115549.2 | 9 | 3213 | |||
XM_011530435.2 | XP_011528737.1 | 10 | 3409 | |||
XM_011530436.2 | XP_011528738.1 | 11 | 3464 | |||
XM_011530437.1 | XP_011528739.1 | 10 | 4134 | |||
XM_011530438.2 | XP_011528740.1 | 10 | 3650 | |||
XM_017028990.1 | XP_016884479.1 | 10 | 3905 | |||
GenBank mRNA | AB051436.1 | - | 9 | 3286 | ||
AK022809.1 | - | 1 | 37 | |||
BC042614.1 | - | 1 | 245 | |||
BC051344.1 | - | 1 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
